Learn More
Cayman Chemical ADB-INACA, Purity: ≥98%, CAS: 1887742-42-8, 1 mg
SDP
Catalog No. 502916466
Encompass_Preferred
An analytical reference standard categorized as a synthetic cannabinoid metabolite and precursor in the synthesis of synthetic cannabinoids; a metabolite of ADB-PINACA and 5-fluoro ADB-PINACA; intended for research and forensic applications
Catalog No. 50-291-6466 Supplier Cayman Chemical Supplier No. 377291 MG
An analytical reference standard categorized as a synthetic cannabinoid metabolite and precursor in the synthesis of synthetic cannabinoids; a metabolite of ADB-PINACA and 5-fluoro ADB-PINACA; intended for research and forensic applications