Learn More
Cayman Chemical AFN-1252, Purity: ≥98%, CAS: 620175-39-5, 25 mg
SDP
Catalog No. 502936585
Encompass_Preferred
An inhibitor of staphylococcal FabI (Ki = 12.8 nM); selective for isolates of methicillin-susceptible and methicillin-resistant S. aureus (MIC90s = ≤0.008 μg/ml) over isolates of S. pneumoniae, β-hemolytic streptococci, Enterococcus, Enterobacteriaceae, non-fermentative Gram-negative bacilli, and M. catarrhalis (MIC90s = >4 μg/ml); increases survival in a murine acute lethal septicemia model at 1 mg/kg
Catalog No. 50-293-6585 Supplier Cayman Chemical Supplier No. 2468625 MG
An inhibitor of staphylococcal FabI (Ki = 12.8 nM); selective for isolates of methicillin-susceptible and methicillin-resistant S. aureus (MIC90s = ≤0.008 μg/ml) over isolates of S. pneumoniae, β-hemolytic streptococci, Enterococcus, Enterobacteriaceae, non-fermentative Gram-negative bacilli, and M. catarrhalis (MIC90s = >4 μg/ml); increases survival in a murine acute lethal septicemia model at 1 mg/kg