Learn More
Cayman Chemical AG-1024, Purity: ≥98%, CAS: 65678-07-1, 10 mg
SDP
Catalog No. 502969879
Encompass_Preferred
A selective inhibitor of IGF-1R that inhibits insulin-stimulated cellular proliferation and IGF-1R autophosphorylation with IC50 values of 0.4 and 7 μM, respectively; inhibits proliferation of human breast cancer MCF-7 cells (10 nM) and human prostate cancer DU145 cells (IC50 = 2.5 μM)
Catalog No. 50-296-9879 Supplier Cayman Chemical Supplier No. 1483310 MG
A selective inhibitor of IGF-1R that inhibits insulin-stimulated cellular proliferation and IGF-1R autophosphorylation with IC50 values of 0.4 and 7 μM, respectively; inhibits proliferation of human breast cancer MCF-7 cells (10 nM) and human prostate cancer DU145 cells (IC50 = 2.5 μM)