Learn More
Cayman Chemical AL 082D06, Purity: ≥98%, CAS: 256925-03-8, 1 mg
SDP
Catalog No. 502987934
Encompass_Preferred
A non-steroidal glucocorticoid receptor antagonist; selective for GR over the androgen, progesterone, mineralocorticoid, and estrogen receptors (Kis = >4, >3, >3, and >10 µM, respectively; inhibits glucocorticoid-induced transcription and reverses dexamethasone-mediated repression of gene expression; inhibits DNA binding in vitro and in vivo; GR bound to AL 082D06 does not compete with a constitutively active GR
Catalog No. 50-298-7934 Supplier Cayman Chemical Supplier No. 234551 MG
A non-steroidal glucocorticoid receptor antagonist; selective for GR over the androgen, progesterone, mineralocorticoid, and estrogen receptors (Kis = >4, >3, >3, and >10 µM, respectively; inhibits glucocorticoid-induced transcription and reverses dexamethasone-mediated repression of gene expression; inhibits DNA binding in vitro and in vivo; GR bound to AL 082D06 does not compete with a constitutively active GR