Learn More
Cayman Chemical Alda-1, Purity: ≥98%, CAS: 349438-38-6, 50 mg
SDP
Catalog No. 502964218
Encompass_Preferred
A selective ALDH2 agonist; enhances wild-type ALDH2*1 and East Asian E487K mutant ALDH2*2 forms of mitochondrial aldehyde dehydrogenase 2 (2.1-, 2.2-, and 11-fold increase in Vmax of 20 µg homozygous WT, heterozygous WT/mutant, and homozygous mutant tetramer, respectively, at 100 µM); has no effect on alcohol dehydrogenase 1, cytosolic ALDH1, or mitochondrial ALDH5; protects against ex vivo ischemic tissue damage to excised rat hearts; reduces infarct size in anesthetized live rats
Catalog No. 50-296-4218 Supplier Cayman Chemical Supplier No. 2155550 MG
A selective ALDH2 agonist; enhances wild-type ALDH2*1 and East Asian E487K mutant ALDH2*2 forms of mitochondrial aldehyde dehydrogenase 2 (2.1-, 2.2-, and 11-fold increase in Vmax of 20 µg homozygous WT, heterozygous WT/mutant, and homozygous mutant tetramer, respectively, at 100 µM); has no effect on alcohol dehydrogenase 1, cytosolic ALDH1, or mitochondrial ALDH5; protects against ex vivo ischemic tissue damage to excised rat hearts; reduces infarct size in anesthetized live rats