Learn More
Cayman Chemical Aloin, Purity: ≥95%, CAS: 452311-56-7, 500 mg
SDP
Catalog No. 502986490
Encompass_Preferred
An anthraquinone with diverse biological activities; reduces IL-6 and TNF-a expression as well as NO production and COX-2 and iNOS expression in LPS-stimulated RAW 264.7 from 100-400 mM; inhibits LPS-induced p65 acetylation, nuclear translocation of NF-kB, and apoptosis in RAW 264.7 cells; reduces motility of T. congolense in vitro and reduces the number of parasites in T. congolense-infected mice at 400 mg/kg; reduces activity of GPX, GST, and GR, the level of colonic MDA, and the number of aberrant crypt foci and mucin-depleted foci in a rat model of 1,2-dimethylhydrazine-induced colon cancer
Catalog No. 50-298-6490 Supplier Cayman Chemical Supplier No. 22435500 MG
An anthraquinone with diverse biological activities; reduces IL-6 and TNF-a expression as well as NO production and COX-2 and iNOS expression in LPS-stimulated RAW 264.7 from 100-400 mM; inhibits LPS-induced p65 acetylation, nuclear translocation of NF-kB, and apoptosis in RAW 264.7 cells; reduces motility of T. congolense in vitro and reduces the number of parasites in T. congolense-infected mice at 400 mg/kg; reduces activity of GPX, GST, and GR, the level of colonic MDA, and the number of aberrant crypt foci and mucin-depleted foci in a rat model of 1,2-dimethylhydrazine-induced colon cancer