Learn More
Cayman Chemical Aloperine, Purity: ≥95%, CAS: 56293-29-9, 1 g
SDP
Catalog No. 502986989
Encompass_Preferred
An alkaloid with diverse biological activities; inhibits HIV-1 replication and envelope-mediated cell-cell fusion (EC50s = 1.75 and 1.2 μM, respectively); inhibits the growth of HL-60, U937, and K562 leukemia cell lines (IC50s = 40, 270, and 360 μM, respectively); reduces NOX2, NOX4, superoxide dismutase, and glutathione peroxidase expression in lungs in a rat model of pulmonary hypertension; reduces ear swelling, ear erythema, and production of inflammatory cytokines TNF-α, IL-1β, and IL-6 in a mouse model of allergic contact dermatitis
Catalog No. 50-298-6989 Supplier Cayman Chemical Supplier No. 222591 G
An alkaloid with diverse biological activities; inhibits HIV-1 replication and envelope-mediated cell-cell fusion (EC50s = 1.75 and 1.2 μM, respectively); inhibits the growth of HL-60, U937, and K562 leukemia cell lines (IC50s = 40, 270, and 360 μM, respectively); reduces NOX2, NOX4, superoxide dismutase, and glutathione peroxidase expression in lungs in a rat model of pulmonary hypertension; reduces ear swelling, ear erythema, and production of inflammatory cytokines TNF-α, IL-1β, and IL-6 in a mouse model of allergic contact dermatitis