Learn More
Cayman Chemical Alphenal, Purity: ≥95%, CAS: 115-43-5, 1 mg
SDP
Catalog No. 502937721
Encompass_Preferred
An analytical reference standard categorized as a barbiturate; intended for research and forensic applications
Catalog No. 50-293-7721 Supplier Cayman Chemical Supplier No. 269181 MG
An analytical reference standard categorized as a barbiturate; intended for research and forensic applications