Learn More
Cayman Chemical AMG 319, Purity: ≥98%, CAS: 1608125-21-8, 5 mg
SDP
Catalog No. 502979689
Encompass_Preferred
A potent, selective inhibitor of PI3Kδ (IC50 = 18 nM); blocks anti-IgM/CD40L-induced proliferation (IC50 = 8.6 nM) and reduces Akt phosphorylation (IC50 = 1.5 nM) in B cells; reduces inflammation in two animal models
Catalog No. 50-297-9689 Supplier Cayman Chemical Supplier No. 173835 MG
A potent, selective inhibitor of PI3Kδ (IC50 = 18 nM); blocks anti-IgM/CD40L-induced proliferation (IC50 = 8.6 nM) and reduces Akt phosphorylation (IC50 = 1.5 nM) in B cells; reduces inflammation in two animal models