Learn More
Cayman Chemical AMG 337, Purity: ≥98%, CAS: 1173699-31-4, 1 mg
SDP
Catalog No. 502985332
Encompass_Preferred
A selective inhibitor of c-Met kinase activity (IC50 = 1 nM) and adaptor protein Gab-1 phosphorylation, which subsequently blocks downstream PI3K and MAPK pathways; inhibits cell proliferation in various c-Met-dependent tumor models
Catalog No. 50-298-5332 Supplier Cayman Chemical Supplier No. 213331 MG
A selective inhibitor of c-Met kinase activity (IC50 = 1 nM) and adaptor protein Gab-1 phosphorylation, which subsequently blocks downstream PI3K and MAPK pathways; inhibits cell proliferation in various c-Met-dependent tumor models