Learn More
Cayman Chemical AMG 517, Purity: ≥98%, CAS: 659730-32-2, 50 mg
SDP
Catalog No. 502935098
Encompass_Preferred
A TRPV1 antagonist (IC50 = 0.9 nM); inhibits mechanical and thermal allodynia in a rat model of burn injury at 165 µg, intrathecally; reverses increases in the level of CGRP and increases expression of GFAP and GAP-43 in the dorsal horn of the spinal cord, as well as enhances axonal regeneration in a rat model of sciatic nerve transection injury
Catalog No. 50-293-5098 Supplier Cayman Chemical Supplier No. 2619150 MG
A TRPV1 antagonist (IC50 = 0.9 nM); inhibits mechanical and thermal allodynia in a rat model of burn injury at 165 µg, intrathecally; reverses increases in the level of CGRP and increases expression of GFAP and GAP-43 in the dorsal horn of the spinal cord, as well as enhances axonal regeneration in a rat model of sciatic nerve transection injury