Learn More
Cayman Chemical AMG 9810, Purity: ≥98%, CAS: 545395-94-6, 25 mg
SDP
Catalog No. 502973321
Encompass_Preferred
A competitive antagonist of capsaicin activation of the TRPV1 (IC50s = 24.5 and 85.6 nM for human and rat, respectively); reverses thermal and mechanical hyperalgesia in a rat model of inflammatory pain
Catalog No. 50-297-3321 Supplier Cayman Chemical Supplier No. 1471525 MG
A competitive antagonist of capsaicin activation of the TRPV1 (IC50s = 24.5 and 85.6 nM for human and rat, respectively); reverses thermal and mechanical hyperalgesia in a rat model of inflammatory pain