Learn More
Cayman Chemical Amlodipine, Purity: ≥98%, CAS: 88150-42-9, 5 g
SDP
Catalog No. 502973075
Encompass_Preferred
An L-type calcium channel blocker; inhibits calcium influx in cardiac and vascular smooth muscle; inhibits calcium-induced contractions in depolarized rat aorta (IC50 = 1.9 nM), displaying a slow rate of association and dissociation in isolated vascular and cardiac tissues
Catalog No. 50-297-3075 Supplier Cayman Chemical Supplier No. 148385 G
An L-type calcium channel blocker; inhibits calcium influx in cardiac and vascular smooth muscle; inhibits calcium-induced contractions in depolarized rat aorta (IC50 = 1.9 nM), displaying a slow rate of association and dissociation in isolated vascular and cardiac tissues