Learn More
Cayman Chemical Amylose, CAS: 9005-82-7, 5 g
SDP
Catalog No. 502966653
Encompass_Preferred
A polysaccharide made of α-D-glucose units that constitutes about 20% of starch; acts as an indicator, turning blue in the presence of iodine, and has also been coupled to magnetic beads for the isolation and purification of maltose-binding protein fusion proteins
Catalog No. 50-296-6653 Supplier Cayman Chemical Supplier No. 176645 G
A polysaccharide made of α-D-glucose units that constitutes about 20% of starch; acts as an indicator, turning blue in the presence of iodine, and has also been coupled to magnetic beads for the isolation and purification of maltose-binding protein fusion proteins