Learn More
BioVendor, LLC Antibody Angiostatin Hu Pab BioVendor LLC
SDP
Supplier:  BioVendor, LLC RD181056100
Encompass
Polyclonal AntibodyWestern blotting, Immunohistochemistry Source of AntigenHuman serumHostsRabbitPreparationThe antibody was raised in rabbits by immunization with the Human Angiostatin from human plasma. The Human Angiostatin contains Kringle 1 ¿ 4 and part of 5.Species ReactivityHuman. Not yet tested in other species.Purification MethodImmunoaffinity chromatography on a column with immobilized serum Human Angiostatin.Antibody Content0.1 mg (determined by BCA method, BSA was used as a standard)
Catalog No. 50-894-027