Learn More
Bioworld Antibody/Antigen Purification Buffer, 1 Kit
Catalog No. 50198580
Encompass_Preferred
Reagents used for binding and eluting inantibody affinitypurification.This kit contains:Tris HCl, pH 8.0 (0.1M, 1L) Glycine, pH 2.5 (0.1M, 500ml)
Catalog No. 50-198-580 Supplier Bioworld Supplier No. 213100011
Reagents used for binding and eluting inantibody affinitypurification.This kit contains:Tris HCl, pH 8.0 (0.1M, 1L) Glycine, pH 2.5 (0.1M, 500ml)