Learn More
Cayman Chemical Apatinib, Purity: ≥98%, CAS: 811803-05-1, 25 mg
SDP
Catalog No. 502985523
Encompass_Preferred
A selective VEGFR2 inhibitor (IC50 = 1 nM); inhibits the proliferation of HUVECs stimulated by FBS and prevents the growth of several established human tumor xenograft models
Catalog No. 50-298-5523 Supplier Cayman Chemical Supplier No. 2126825 MG
A selective VEGFR2 inhibitor (IC50 = 1 nM); inhibits the proliferation of HUVECs stimulated by FBS and prevents the growth of several established human tumor xenograft models