Learn More
Cayman Chemical AR-7, Purity: ≥98%, CAS: 80306-38-3, 1 mg
SDP
Catalog No. 502995465
Encompass_Preferred
A RARα antagonist; increases chaperone-mediated autophagy in an RARα-dependent manner, increases LAMP-2A expression, and inhibits paraquat-induced decreases in cell viability in mouse fibroblasts at 20 μM; selectively cytotoxic to HepG2 cells grown under hypoxic over normoxic conditions (IC50s = 10 and >1,000 μM, respectively)
Catalog No. 50-299-5465 Supplier Cayman Chemical Supplier No. 229461 MG
A RARα antagonist; increases chaperone-mediated autophagy in an RARα-dependent manner, increases LAMP-2A expression, and inhibits paraquat-induced decreases in cell viability in mouse fibroblasts at 20 μM; selectively cytotoxic to HepG2 cells grown under hypoxic over normoxic conditions (IC50s = 10 and >1,000 μM, respectively)