Learn More
Cayman Chemical Arctiin, Purity: ≥98%, CAS: 20362-31-6, 25 mg
SDP
Catalog No. 502972450
Encompass_Preferred
The major active lignin in fruits of the burdock plant A. lappa that demonstrates potent antiviral, anti-inflammatory, and antiproliferative activity
Catalog No. 50-297-2450 Supplier Cayman Chemical Supplier No. 1537525 MG
The major active lignin in fruits of the burdock plant A. lappa that demonstrates potent antiviral, anti-inflammatory, and antiproliferative activity