Learn More
Cayman Chemical ARN24139, Purity: ≥98%, CAS: 2699768-78-8, 10 mg
SDP
Catalog No. 502988412
Encompass_Preferred
A topoisomerase II poison (IC50 = 7.3 µM); inhibits proliferation of DU145, HeLa, and A549 cells (IC50s = 4.7, 3.8, and 3.1 µM, respectively)
Catalog No. 50-298-8412 Supplier Cayman Chemical Supplier No. 3106210 MG
A topoisomerase II poison (IC50 = 7.3 µM); inhibits proliferation of DU145, HeLa, and A549 cells (IC50s = 4.7, 3.8, and 3.1 µM, respectively)