Learn More
Description
- For best results, store the components as described in the protocol
- Avoid repeated freezing and thawing
Specifications
Specifications
| For Use With (Application) | Profiling |
| Product Type | Array/Library |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Quantity | 1 Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.