Learn More
Raybiotech Inc C-Series Gallus (Chicken) Cytokine Array C1 Kit.
Catalog No. 501948452
Encompass_Preferred
Detects 10 Gallus Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Caronte, IFN-gamma, IL-6, IL-10, IL-12 p40, IL-16, IL-21, Netrin-2, Pentraxin-3, RANTES
Catalog No. 50-194-8452 Supplier Raybiotech Inc Supplier No. AAGCYT12
Add to Cart
Add to Cart
Detects 10 Gallus Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Caronte, IFN-gamma, IL-6, IL-10, IL-12 p40, IL-16, IL-21, Netrin-2, Pentraxin-3, RANTES