Learn More
Raybiotech Inc C-Series Human and Mouse AKT Pathway Phosphorylation Array C1.
Catalog No. 501948459
Encompass_Preferred
Detects 18 phosphorylated human and mouse proteins. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Akt (P-Ser473) (PKB (P-Ser473)), AMPKa (P-Thr172), BAD (P-Ser112), 4E-BP1 (P-Thr36), ERK1 (P-T202/Y204)/ERK2 (P-Y185/Y187), GSK3a (P-Ser21), GSK3b (P-Ser9), mTOR (P-Ser2448), p27 (P-Thr198), P53 (P-Ser15), P70S6K (P-Thr421/Ser424), PDK1 (P-S241), PRAS40 (P-Thr246), PTEN (P-Ser380), Raf-1 (Ser301), RPS6 (P-Ser235/Ser236), RSK1 (P-Ser380), RSK2 (P-Ser386)
Catalog No. 50-194-8459 Supplier Raybiotech Inc Supplier No. AAHAKT12
Detects 18 phosphorylated human and mouse proteins. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Akt (P-Ser473) (PKB (P-Ser473)), AMPKa (P-Thr172), BAD (P-Ser112), 4E-BP1 (P-Thr36), ERK1 (P-T202/Y204)/ERK2 (P-Y185/Y187), GSK3a (P-Ser21), GSK3b (P-Ser9), mTOR (P-Ser2448), p27 (P-Thr198), P53 (P-Ser15), P70S6K (P-Thr421/Ser424), PDK1 (P-S241), PRAS40 (P-Thr246), PTEN (P-Ser380), Raf-1 (Ser301), RPS6 (P-Ser235/Ser236), RSK1 (P-Ser380), RSK2 (P-Ser386)