Learn More
Raybiotech Inc Quantibody Human Adhesion Molecule Array 1 Kit.
Catalog No. 501950811
Encompass_Preferred
Detects detects 17 human cell adhesion molecules. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: ALCAM (CD166), BCAM, CEACAM-1, E-Cadherin, TROP1 (EpCAM), E-Selectin, ICAM-1 (CD54), ICAM-2 (CD102), ICAM-3 (CD50), L-Selectin (CD62L), NCAM-1 (CD56), NrCAM, P-Cadherin, P-Selectin, PECAM-1 (CD31), VCAM-1 (CD106), VE-Cadherin (CDH5)
Catalog No. 50-195-0811 Supplier Raybiotech Inc Supplier No. QAHCAM11
Detects detects 17 human cell adhesion molecules. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: ALCAM (CD166), BCAM, CEACAM-1, E-Cadherin, TROP1 (EpCAM), E-Selectin, ICAM-1 (CD54), ICAM-2 (CD102), ICAM-3 (CD50), L-Selectin (CD62L), NCAM-1 (CD56), NrCAM, P-Cadherin, P-Selectin, PECAM-1 (CD31), VCAM-1 (CD106), VE-Cadherin (CDH5)