Learn More
Raybiotech Inc Quantibody Human IL-1 Family Cytokine Array 1 Kit.
Catalog No. 501950838
Encompass_Preferred
Detects all the 11 IL-1 Family Cytokine members. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-1 RA (IL-1 F3), IL-18, IL-33 (IL-1 F11), IL-36 alpha, IL-36 beta, IL-36 gamma, IL-36 RA, IL-37, IL-38
Catalog No. 50-195-0838 Supplier Raybiotech Inc Supplier No. QAHIL1F12
Detects all the 11 IL-1 Family Cytokine members. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-1 RA (IL-1 F3), IL-18, IL-33 (IL-1 F11), IL-36 alpha, IL-36 beta, IL-36 gamma, IL-36 RA, IL-37, IL-38