Learn More
Raybiotech Inc Quantibody Human Periodontal Disease Array 1 Kit.
Catalog No. 501950852
Encompass_Preferred
Detects 20 Human Cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: CRP (C-Reactive Protein), IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-10, IL-12 p70, IL-17A, IL-2, IL-4, IL-6, IL-8 (CXCL8), MIP-1 alpha (CCL3), MMP-13, MMP-9, Osteoactivin (GPNMB), Osteopontin (SPP1), Osteoprotegerin (TNFRSF11B), RANK (TNFRSF11A), TGF beta 1, TNF alpha
Catalog No. 50-195-0852 Supplier Raybiotech Inc Supplier No. QAHPDD12
Detects 20 Human Cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: CRP (C-Reactive Protein), IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-10, IL-12 p70, IL-17A, IL-2, IL-4, IL-6, IL-8 (CXCL8), MIP-1 alpha (CCL3), MMP-13, MMP-9, Osteoactivin (GPNMB), Osteopontin (SPP1), Osteoprotegerin (TNFRSF11B), RANK (TNFRSF11A), TGF beta 1, TNF alpha