Learn More
Raybiotech Inc Quantibody Non-Human Primate Cytokine Array 1 Kit.
Supplier:  Raybiotech Inc QANCYT11
Encompass_Preferred
Detects 10 Non-Human Primate Cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: GM-CSF, IFN-gamma, IL-1 beta (IL-1 F2), IL-4, IL-5, IL-6, IL-12 p70, IL-15, IL-16, TNF alpha
Catalog No. 50-195-0897