Learn More
Raybiotech Inc C-Series Human Immune Checkpoint Antibody Array 1 Kit.
Supplier:  Raybiotech Inc AAHICM14
Encompass_Preferred
Detects 23 Human molecules expressed in T/B cells and APCs. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: 4-1BB (TNFRSF9/CD137), B7-H2 (ICOS Ligand), B7-H3 (CD276), B7-H4, CD200, CD200 R1 CD27 (TNFRSF7), HVEM (TNFRSF14/CD270), CD27 Ligand (TNFSF7), CD28, CD40 Ligand (TNFSF5), CD80 (B7-1), CD86 (B7-2), CTLA-4 (CD152), ICOS, LAG-3, OX40 (TNFRSF4), OX40 Ligand (TNFSF4), PD-1, B7-H1 (CD274/PD-L1), PD-L2 (B7-DC), TIM-1 (KIM-1), TIM-3 (KIM-3)
Catalog No. 50-194-8592