Learn More
Raybiotech Inc C-Series Human Immune Checkpoint Antibody Array 2 Kit.
Supplier: Raybiotech Inc AAHICM28
Detects 11 Human molecules expressed in T/B cells. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: CD200 R1, CD27 (TNFRSF7), CD28, CD40 Ligand (TNFSF5), CTLA-4 (CD152), ICOS, LAG-3, OX40 (TNFRSF4), PD-1, TIM-1 (KIM-1), TIM-3 (KIM-3)