Learn More
Raybiotech Inc C-Series Human Insulin Receptor Pathway Phosphorylation Array 1 Kit.
Catalog No. 501948612
Encompass_Preferred
Detects 9 phosphorylated proteins in human samples. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: eIF-4E, FOXO-3, IGF-1 R, Insulin, IRS-1, LKB1, SHC, SHIP1, SHP-2
Catalog No. 50-194-8612 Supplier Raybiotech Inc Supplier No. AAHINSR14
Detects 9 phosphorylated proteins in human samples. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: eIF-4E, FOXO-3, IGF-1 R, Insulin, IRS-1, LKB1, SHC, SHIP1, SHP-2