Learn More
Raybiotech Inc C-Series Human JAK/STAT Pathway Phosphorylation Array 1 Kit.
Supplier:  Raybiotech Inc AAHJAKSTAT18
Encompass_Preferred
Detects 12 phosphorylated or cleaved human Apoptotic Factors. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: EGFR (P-Ser1070), JAK1 (P-Tyr1022), JAK2 (P-Tyr1007/1008), SHP-1 (P-Ser591), SHP2 (P-Tyr542), Src (P-Tyr419), STAT1 (P-Ser727), STAT2 (P-Tyr689), STAT3 (P-Tyr705), STAT5 (P-Tyr694), STAT6 (P-Tyr641), TYK2 (P-Tyr1054)
Catalog No. 50-194-8616