Learn More
Raybiotech Inc C-Series Human/Mouse MAPK Phosphorylation Antibody Array 1 Kit.
Catalog No. 501948619
Encompass_Preferred
Detects 17 Human/Mouse proteins. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Akt (P-Ser473), CREB (P-Ser133), ERK1 (P-T202/Y204)/ERK2 (P-Y185/Y187), GSK3a (P-Ser21), GSK3b (P-Ser9), HSP27 (P-Ser82), JNK (P-Thr183), MEK (P-Ser217/Ser221), MKK3 (P-Ser189), MKK6 (P-Ser207), MSK2 (P-Ser360), mTOR (P-Ser2448), p38 (P-Thr180/Tyr182), P53 (P-Ser15), P70S6K (P-Thr421/Ser424), RSK1 (P-Ser380), RSK2 (P-Ser386)
Catalog No. 50-194-8619 Supplier Raybiotech Inc Supplier No. AAHMAPK18
Detects 17 Human/Mouse proteins. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Akt (P-Ser473), CREB (P-Ser133), ERK1 (P-T202/Y204)/ERK2 (P-Y185/Y187), GSK3a (P-Ser21), GSK3b (P-Ser9), HSP27 (P-Ser82), JNK (P-Thr183), MEK (P-Ser217/Ser221), MKK3 (P-Ser189), MKK6 (P-Ser207), MSK2 (P-Ser360), mTOR (P-Ser2448), p38 (P-Thr180/Tyr182), P53 (P-Ser15), P70S6K (P-Thr421/Ser424), RSK1 (P-Ser380), RSK2 (P-Ser386)