Learn More
Raybiotech Inc C-Series Human JAK/STAT Pathway Phosphorylation Array 1 Kit.
Catalog No. 501948614
Encompass_Preferred
Detects 12 phosphorylated or cleaved human Apoptotic Factors. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: EGFR (P-Ser1070), JAK1 (P-Tyr1022), JAK2 (P-Tyr1007/1008), SHP-1 (P-Ser591), SHP2 (P-Tyr542), Src (P-Tyr419), STAT1 (P-Ser727), STAT2 (P-Tyr689), STAT3 (P-Tyr705), STAT5 (P-Tyr694), STAT6 (P-Tyr641), TYK2 (P-Tyr1054)
Catalog No. 50-194-8614 Supplier Raybiotech Inc Supplier No. AAHJAKSTAT12
Detects 12 phosphorylated or cleaved human Apoptotic Factors. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: EGFR (P-Ser1070), JAK1 (P-Tyr1022), JAK2 (P-Tyr1007/1008), SHP-1 (P-Ser591), SHP2 (P-Tyr542), Src (P-Tyr419), STAT1 (P-Ser727), STAT2 (P-Tyr689), STAT3 (P-Tyr705), STAT5 (P-Tyr694), STAT6 (P-Tyr641), TYK2 (P-Tyr1054)