Learn More
Raybiotech Inc C-Series Equine (Horse) Cytokine Array C1 Kit.
Catalog No. 501948451
Encompass_Preferred
Detects 10 Equine Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: IFN-gamma, IL-1 alpha, IL-1 Ra, IL-2, IL-4, IL-8, IL-10, IL-15, MCP-1, VEGF-A
Catalog No. 50-194-8451 Supplier Raybiotech Inc Supplier No. AAECYT12
Detects 10 Equine Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: IFN-gamma, IL-1 alpha, IL-1 Ra, IL-2, IL-4, IL-8, IL-10, IL-15, MCP-1, VEGF-A