Learn More
Raybiotech Inc C-Series Human Phosphorylation Pathway Profiling Array C55 Kit.
Catalog No. 501948639
Encompass_Preferred
Detects phosphorylated human proteins in 5 signaling pathways: MAPK, AKT, JAK/STAT, NFκB, and TGFβ. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. View protein list on RayBiotech’s website – AAH-PPP-1-4
Catalog No. 50-194-8639 Supplier Raybiotech Inc Supplier No. AAHPPP14
Detects phosphorylated human proteins in 5 signaling pathways: MAPK, AKT, JAK/STAT, NFκB, and TGFβ. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. View protein list on RayBiotech’s website – AAH-PPP-1-4