Learn More
Raybiotech Inc G-Series Human Receptor Tyrosine Kinase Phosphorylation Antibody Array 1 Kit.
Catalog No. 501948644
Encompass_Preferred
Detects 71 Human RTKs. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: ABL1, ACK, ALK-1, Axl, Blk, BMX, Btk, Csk, Dtk, EGFR, EphA1, EphA2, EphA3, EphA4, EphA5, EphA6, EphA7, EphA8, EphB1, EphB2, EphB3, EphB4, EphB6, ErbB2, ErbB3, ErbB4, FAK, FER, FGFR1, FGFR2, FGFR2 (alpha isoform), Fgr, FRK, Fyn, Hck, HGFR, IGF-1 R, Insulin R (CD220), Itk, JAK1, JAK2, JAK3, LCK, LTK, Lyn, MATK, M-CSF R, MUSK, NGFR (TNFRSF16), PDGFRA, PDGFRB, PYK2, RET, ROR1, ROR2, ROS, RYK, SCF R (CD117/c-kit), SRMS, SYK, Tec, Tie-1, Tie-2, TNK1, TRKB, TXK, Tyk2, TYRO10 (DDR2/TKT), VEGFR2, VEGFR3
Catalog No. 50-194-8644 Supplier Raybiotech Inc Supplier No. AAHPRTKG14
Detects 71 Human RTKs. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: ABL1, ACK, ALK-1, Axl, Blk, BMX, Btk, Csk, Dtk, EGFR, EphA1, EphA2, EphA3, EphA4, EphA5, EphA6, EphA7, EphA8, EphB1, EphB2, EphB3, EphB4, EphB6, ErbB2, ErbB3, ErbB4, FAK, FER, FGFR1, FGFR2, FGFR2 (alpha isoform), Fgr, FRK, Fyn, Hck, HGFR, IGF-1 R, Insulin R (CD220), Itk, JAK1, JAK2, JAK3, LCK, LTK, Lyn, MATK, M-CSF R, MUSK, NGFR (TNFRSF16), PDGFRA, PDGFRB, PYK2, RET, ROR1, ROR2, ROS, RYK, SCF R (CD117/c-kit), SRMS, SYK, Tec, Tie-1, Tie-2, TNK1, TRKB, TXK, Tyk2, TYRO10 (DDR2/TKT), VEGFR2, VEGFR3