Learn More
Raybiotech Inc C-Series Human NF-kB Pathway Phosphorylation Array 1 Kit.
Catalog No. 501948631
Encompass_Preferred
Detects 11 phosphorylated proteins in human samples. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: ATM (P-Ser1981), eIF-2a (P-Ser52), HDAC2 (P-Ser394), HDAC4 (P-Ser632), IKB-alpha (P-Ser32), MSK1 (P-Ser376), NF-kB (P-Ser536), STAT1 (P-Ser727), TAK1 (P-Ser412), TBK1 (P-Ser172), ZAP70 (P-Tyr292)
Catalog No. 50-194-8631 Supplier Raybiotech Inc Supplier No. AAHNFKB18
Detects 11 phosphorylated proteins in human samples. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: ATM (P-Ser1981), eIF-2a (P-Ser52), HDAC2 (P-Ser394), HDAC4 (P-Ser632), IKB-alpha (P-Ser32), MSK1 (P-Ser376), NF-kB (P-Ser536), STAT1 (P-Ser727), TAK1 (P-Ser412), TBK1 (P-Ser172), ZAP70 (P-Tyr292)