Learn More
Raybiotech Inc C-Series Human EGFR Phosphorylation Antibody Array 1 Kit.
Catalog No. 501948637
Encompass_Preferred
Detects 17 site specific and pan Human EGFRs. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: EGFR (P-Tyr845), EGFR (P-Tyr992), EGFR (P-Tyr1045), EGFR (P-Tyr1068), EGFR (P-Tyr1086), EGFR (P-Tyr1148), EGFR (P-Tyr1173), EGFR (P-Ser1046/1047), EGFR (P-Ser1070), ErbB2 (P-Tyr877), ErbB2 (P-Tyr1112), ErbB2 (P-Tyr 1221/1222), ErbB2 (P-Tyr1248), ErbB2 (P-Thr686), ErbB2 (P-Ser1113), ErbB3 (P-Tyr1289), ErbB4 (P-Tyr1284)
Catalog No. 50-194-8637 Supplier Raybiotech Inc Supplier No. AAHPER18
Detects 17 site specific and pan Human EGFRs. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: EGFR (P-Tyr845), EGFR (P-Tyr992), EGFR (P-Tyr1045), EGFR (P-Tyr1068), EGFR (P-Tyr1086), EGFR (P-Tyr1148), EGFR (P-Tyr1173), EGFR (P-Ser1046/1047), EGFR (P-Ser1070), ErbB2 (P-Tyr877), ErbB2 (P-Tyr1112), ErbB2 (P-Tyr 1221/1222), ErbB2 (P-Tyr1248), ErbB2 (P-Thr686), ErbB2 (P-Ser1113), ErbB3 (P-Tyr1289), ErbB4 (P-Tyr1284)