Learn More
Raybiotech Inc C-Series Human TGF beta Pathway Phosphorylation Array 1 Kit.
Catalog No. 501948661
Encompass_Preferred
Detects 8 phosphorylated proteins in human samples. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: c-Jun, SMAD1, SMAD5, SMAD4, ATF2, C-Fos, SMAD2, TAK2
Catalog No. 50-194-8661 Supplier Raybiotech Inc Supplier No. AAHTGFB14
Detects 8 phosphorylated proteins in human samples. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: c-Jun, SMAD1, SMAD5, SMAD4, ATF2, C-Fos, SMAD2, TAK2