Learn More
Raybiotech Inc C-Series Mouse Apoptosis Antibody Array 1 Kit.
Supplier:  Raybiotech Inc AAMAPOSIG12
Encompass_Preferred
Detects 17 phosphorylated or cleaved mouse Apoptotic Factors. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: AKT (P-Ser473), ATM (P-Ser1981), BAD (P-Ser112), Caspase-3 (P-Asp175), Caspase-7 (P-Asp198), CHK1 (P-Ser296), eIF-2a (P-Ser52), ERK1/2 (P-Thr202), HSP27 (P-Ser82), IKBa (P-Ser32), JNK (P-Thr183), NF-kB P65 (P-Ser536), p27 (P-Thr198), p38 (P-Thr180/Tyr182), P53 (P-Ser15), SMAD2 (P-Ser245), TAK1 (P-Ser412)
Catalog No. 50-194-8674