Learn More
Raybiotech Inc C-Series Mouse Apoptosis Antibody Array 1 Kit.
Catalog No. 501948675
Encompass_Preferred
Detects 17 phosphorylated or cleaved mouse Apoptotic Factors. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: AKT (P-Ser473), ATM (P-Ser1981), BAD (P-Ser112), Caspase-3 (P-Asp175), Caspase-7 (P-Asp198), CHK1 (P-Ser296), eIF-2a (P-Ser52), ERK1/2 (P-Thr202), HSP27 (P-Ser82), IKBa (P-Ser32), JNK (P-Thr183), NF-kB P65 (P-Ser536), p27 (P-Thr198), p38 (P-Thr180/Tyr182), P53 (P-Ser15), SMAD2 (P-Ser245), TAK1 (P-Ser412)
Catalog No. 50-194-8675 Supplier Raybiotech Inc Supplier No. AAMAPOSIG14
Detects 17 phosphorylated or cleaved mouse Apoptotic Factors. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: AKT (P-Ser473), ATM (P-Ser1981), BAD (P-Ser112), Caspase-3 (P-Asp175), Caspase-7 (P-Asp198), CHK1 (P-Ser296), eIF-2a (P-Ser52), ERK1/2 (P-Thr202), HSP27 (P-Ser82), IKBa (P-Ser32), JNK (P-Thr183), NF-kB P65 (P-Ser536), p27 (P-Thr198), p38 (P-Thr180/Tyr182), P53 (P-Ser15), SMAD2 (P-Ser245), TAK1 (P-Ser412)