Learn More
Raybiotech Inc C-Series Mouse Immune Checkpoint Antibody Array 1 Kit.
Supplier:  Raybiotech Inc AAMICM12
Encompass_Preferred
Detects 11 mouse immune checkpoints. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: 4-1BB (TNFRSF9/CD137),CD27 (TNFRSF7), CD40 (TNFRSF5), CD40 Ligand (TNFSF5), CD80 (B7-1), CTLA-4 (CD152), Fas (TNFRSF6/Apo-1), Fas Ligand (TNFSF6), Galectin-1, Galectin-3, OX40 Ligand (TNFSF4)
Catalog No. 50-194-8737