Learn More
Raybiotech Inc C-Series Ovine (Sheep) Cytokine Array C1 Kit.
Supplier:  Raybiotech Inc AAOCYT14
Encompass_Preferred
Detects 18 Ovine Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: AIF, Cadherin-6, Decorin, IFN alpha, IFN-gamma, IL-1 alpha, IL-1 beta, IL-4, IL-13, IL-17A, IL-21, LEKTI, LIF, MIG, RANTES, sFRP-3, TNF alpha, VEGF-A
Catalog No. 50-194-8747