Learn More
Raybiotech Inc C-Series Porcine T Cell Response Array C1 Kit.
Supplier:  Raybiotech Inc AAPTCR14
Encompass_Preferred
Detects 19 Porcine T Cell Response factors. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: GM-CSF, IFN-gamma, IL-1 alpha, IL-1 beta, IL-1 Ra, IL-10, IL-12 p70, IL-13, IL-15, IL-17A, IL-17F, IL-21, IL-22, IL-28B, IL-4, IL-6, NCAM-1, TGF beta 1, TNF alpha
Catalog No. 50-194-8762