Learn More
Raybiotech Inc C-Series Rat Immune Checkpoint Molecule Array 1 Kit.
Supplier:  Raybiotech Inc AARICM12
Encompass_Preferred
Detects 9 rat Immune Checkpoints. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: 4-1BB, CD27, CD80, CD86, Galectin-1, Galectin-3, Fas, Fas Ligand, TIM-1
Catalog No. 50-194-8787