Learn More
Raybiotech Inc G-Series Human Bone Metabolism Array 2 Kit.
Catalog No. 501950369
Encompass_Preferred
Semi-quantitative measurement of 10 human bone metabolism associated cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: BMP-2, BMP-6, BMP-7, DKK-1, MMP-3, Osteoprotegerin (TNFRSF11B), Osteopontin (SPP1), PDGF-BB, TGF beta 3, TRANCE (TNFSF11)
Catalog No. 50-195-0369 Supplier Raybiotech Inc Supplier No. GSHBMA22
Semi-quantitative measurement of 10 human bone metabolism associated cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: BMP-2, BMP-6, BMP-7, DKK-1, MMP-3, Osteoprotegerin (TNFRSF11B), Osteopontin (SPP1), PDGF-BB, TGF beta 3, TRANCE (TNFSF11)