Learn More
Raybiotech Inc G-Series Human Bone Metabolism Array 1 Kit.
Supplier:  Raybiotech Inc GSHBMA12
Encompass_Preferred
Semi-quantitative measurement of 31 human bone metabolism associated cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Activin A, aFGF (FGF-1), Amphiregulin, bFGF, BMP-4, BMP-9, E-Selectin, ICAM-1 (CD54), IGF-1, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-6, IL-8 (CXCL8), IL-11, IL-17A, MCP-1 (CCL2), M-CSF, MIP-1 alpha (CCL3), MMP-2, MMP-9, MMP-13, Osteoactivin (GPNMB), P-Cadherin, RANK (TNFRSF11A), SDF-1 alpha (CXCL12 alpha), Sonic Hedgehog N-Terminal (Shh-N), TGF beta 1, TGF beta 2, TNF alpha, VCAM-1 (CD106), VE-Cadherin (CDH5)
Catalog No. 50-195-0367