Learn More
Raybiotech Inc G-Series Human Serum Abundant Protein Array 2 Kit.
Supplier:  Raybiotech Inc GSHSAP21
Encompass_Preferred
Semi-quantitative measurement of 31 Human Serum Abundant serum proteins. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Adipsin (Complement Factor D), Angiogenin, beta IG-H3, Beta-2 Microglobulin, CD14, CD26 (DPPIV), CRP (C-Reactive Protein), Endostatin, E-Selectin, FAP, Ferritin, Fetuin A, gp130 (sgp130), HGFR, ICAM-2 (CD102), IGFBP-2, IGFBP-3, IGFBP-6, Lactoferrin, Lipocalin-2 (NGAL), L-Selectin (CD62L), LYVE-1, M-CSF R, MMP-9, NCAM-1 (CD56), Nidogen-1, PAI-1, Platelet Factor 4 (CXCL4), RBP4, SAA (Serum Amyloid A), Thrombospondin 1
Catalog No. 50-195-0422