Learn More
Raybiotech Inc G-Series Human Serum Abundant Protein Array 3 Kit.
Supplier:  Raybiotech Inc GSHSAP32
Encompass_Preferred
Semi-quantitative measurement of 31 Human Serum Abundant serum proteins. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Adiponectin (ACRP30), Adipsin (Complement Factor D), Albumin, Alpha-2 Macroglobulin, Angiogenin, ApoA, ApoC1, ApoC2, ApoC3, ApoE, ApoH, Beta-2 Microglobulin, CD14, Clusterin, CRP (C-Reactive Protein), Cystatin C, ECM-1, Fetuin A, Lipocalin-2 (NGAL), LYVE-1, MBL, NAP-2 (PPBP/CXCL7), Platelet Factor 4 (CXCL4), RBP4, SAA (Serum Amyloid A), SERPINA4, TfR (Transferrin R), Thrombospondin 1, Transferrin, Vitronectin, VWF
Catalog No. 50-195-0425