Learn More
Raybiotech Inc G-Series Human Adhesion Molecule Array 1 Kit.
Supplier:  Raybiotech Inc GSHCAM12
Encompass_Preferred
Semi-quantitative measurement of 17 human cell adhesion molecules. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: ALCAM (CD166), BCAM, CEACAM-1, E-Cadherin, TROP1 (EpCAM), E-Selectin, ICAM-1 (CD54), ICAM-2 (CD102), ICAM-3 (CD50), L-Selectin (CD62L), NCAM-1 (CD56), NrCAM, P-Cadherin, P-Selectin, PECAM-1 (CD31), VCAM-1 (CD106), VE-Cadherin (CDH5)
Catalog No. 50-195-0379